Anti-IgA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Abcam
ABCAAB8510-500EA 248.99 CHF
ABCAAB8510-500
Anti-IgA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR