Anti-MMP3 Sheep Polyclonal Antibody

ABCAAB8482-1EA 660 CHF
ABCAAB8482-1
Anti-MMP3 Sheep Polyclonal Antibody
Type: Primary
Antigen: MMP3 (matrix metallopeptidase 3 (stromelysin 1, progelatinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR