Anti-SPP1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB8448-200EA 560.23 CHF
ABCAAB8448-200
Anti-SPP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SPP1 (secreted phosphoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR