Anti-RIPK2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB8428-100EA 492.8 CHF
ABCAAB8428-100
Anti-RIPK2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RIPK2 (receptor-interacting serine-threonine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR