Anti-CASP8AP2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB8420-100EA 508.36 CHF
ABCAAB8420-100
Anti-CASP8AP2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CASP8AP2 (caspase 8 associated protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR