Anti-TNFRSF10A Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB8414-100EA 951 CHF
ABCAAB8414-100
Anti-TNFRSF10A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNFRSF10A (tumor necrosis factor receptor superfamily, member 10a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR