Anti-DFFB Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB8408-100EA 508.36 CHF
ABCAAB8408-100
Anti-DFFB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DFFB (DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR