Anti-TNFRSF6B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB8405-100EA 524.65 CHF
ABCAAB8405-100
Anti-TNFRSF6B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TNFRSF6B (tumor necrosis factor receptor superfamily, member 6b, decoy)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR