Anti-Ovalbumin Rabbit Polyclonal Antibody Biotin

ABCAAB8389-1EA 672 CHF
ABCAAB8389-1
Anti-Ovalbumin Rabbit Polyclonal Antibody Biotin
Type: Primary
Antigen: Ovalbumin
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR