Anti-GAPDH Mouse Monoclonal Antibody [clone: 6C5]

Supplier: Abcam
ABCAAB8245-100EA 612.11 CHF
ABCAAB8245-100
Anti-GAPDH Mouse Monoclonal Antibody [clone: 6C5]
Antibodies
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: monoclonal
Clone: 6C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR