Anti-IL2RA Mouse Monoclonal Antibody [clone: MEM-181]

Supplier: Abcam
ABCAAB8235-100EA 430.55 CHF
ABCAAB8235-100
Anti-IL2RA Mouse Monoclonal Antibody [clone: MEM-181]
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone: MEM-181
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR