Anti-CD63 Mouse Monoclonal Antibody [clone: MEM-259]

Supplier: Abcam
ABCAAB8219-100EA 472.05 CHF
ABCAAB8219-100
Anti-CD63 Mouse Monoclonal Antibody [clone: MEM-259]
Antibodies
Type: Primary
Antigen: CD63 (CD63 molecule)
Clonality: monoclonal
Clone: MEM-259
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR