Anti-IL1RL1 Rat Monoclonal Antibody [clone: REG21]
Lieferant: Abcam
ABCAAB8154-500EA 505.57 CHF
ABCAAB8154-500
Anti-IL1RL1 Rat Monoclonal Antibody [clone: REG21]
Antikörper
Type: Primary
Antigen: IL1RL1 (interleukin 1 receptor-like 1)
Clonality: monoclonal
Clone: REG21
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse