Anti-200 kD Neurofilament Heavy Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB8135-50EA 586.17 CHF
ABCAAB8135-50
Anti-200 kD Neurofilament Heavy Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: 200 kD Neurofilament Heavy
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR