Anti-IRAKM Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB8116-100EA 529.11 CHF
ABCAAB8116-100
Anti-IRAKM Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRAKM
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR