Anti-CHEK2 Rabbit Polyclonal Antibody

ABCAAB8108-100EA 749 CHF
ABCAAB8108-100
Anti-CHEK2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR