Anti-Mannose 6 Phosphate Receptor (Cation independent) Mouse Monoclonal Antibody [clone: MEM-238]

Supplier: Abcam
ABCAAB8093-100EA 456.49 CHF
ABCAAB8093-100
Anti-Mannose 6 Phosphate Receptor (Cation independent) Mouse Monoclonal Antibody [clone: MEM-238]
Antibodies
Type: Primary
Antigen: Mannose 6 Phosphate Receptor (Cation independent)
Clonality: monoclonal
Clone: MEM-238
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR