Anti-Campylobacter jejuni Mouse Monoclonal Antibody [clone: 57-24]
Lieferant: Abcam
ABCAAB8063-1EA 524.65 CHF
ABCAAB8063-1
Anti-Campylobacter jejuni Mouse Monoclonal Antibody [clone: 57-24]
Antikörper
Type: Primary
Antigen: Campylobacter jejuni
Clonality: monoclonal
Clone: 57-24
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now