Anti-SAA@ Mouse Monoclonal Antibody [clone: Reu-86.2]

Supplier: Abcam
ABCAAB8057-500EA 492.8 CHF
ABCAAB8057-500
Anti-SAA@ Mouse Monoclonal Antibody [clone: Reu-86.2]
Antibodies
Type: Primary
Antigen: SAA@ (serum amyloid A1 cluster)
Clonality: monoclonal
Clone: Reu-86.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR