Anti-BCL2L1 Rabbit Polyclonal Antibody

ABCAAB7974-1EA 708 CHF
ABCAAB7974-1
Anti-BCL2L1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: BCL2L1 (BCL2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR