Anti-Hepatitis B Virus Core Antigen Rabbit Polyclonal Antibody

ABCAAB7841-500EA 699 CHF
ABCAAB7841-500
Anti-Hepatitis B Virus Core Antigen Rabbit Polyclonal Antibody
Type: Primary
Antigen: Hepatitis B Virus Core Antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR