Anti-liver FABP Rabbit Polyclonal Antibody

ABCAAB7807-500EA 725 CHF
ABCAAB7807-500
Anti-liver FABP Rabbit Polyclonal Antibody
Type: Primary
Antigen: liver FABP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR