Anti-HLA G Mouse Monoclonal Antibody [clone: MEM-G/1]

Supplier: Abcam
ABCAAB7759-100EA 456.49 CHF
ABCAAB7759-100
Anti-HLA G Mouse Monoclonal Antibody [clone: MEM-G/1]
Antibodies
Type: Primary
Antigen: HLA G
Clonality: monoclonal
Clone: MEM-G/1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR