Anti-APOC1 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB7617-100EA 903 CHF
ABCAAB7617-100
Anti-APOC1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APOC1 (apolipoprotein C-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR