Anti-EPCAM Mouse Monoclonal Antibody [clone: Ber-EP4]

Supplier: Abcam
ABCAAB7504-500EA 549.86 CHF
ABCAAB7504-500
Anti-EPCAM Mouse Monoclonal Antibody [clone: Ber-EP4]
Antibodies
Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: monoclonal
Clone: Ber-EP4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR