Anti-ACHE Goat Polyclonal Antibody

ABCAAB7396-1EA 672 CHF
ABCAAB7396-1
Anti-ACHE Goat Polyclonal Antibody
Type: Primary
Antigen: ACHE (acetylcholinesterase (Yt blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR