Anti-CSF2 Rat Monoclonal Antibody [clone: MP1-22E9]

ABCAAB7368-50EA 672 CHF
ABCAAB7368-50
Anti-CSF2 Rat Monoclonal Antibody [clone: MP1-22E9]
Type: Primary
Antigen: CSF2 (colony stimulating factor 2 (granulocyte-macrophage))
Clonality: monoclonal
Clone: MP1-22E9
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR