Anti-pan methyl Lysine Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB7315-50EA 457.88 CHF
ABCAAB7315-50
Anti-pan methyl Lysine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: pan methyl Lysine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now