Anti-PSEN2 Goat Polyclonal Antibody

ABCAAB7301-50EA 699 CHF
ABCAAB7301-50
Anti-PSEN2 Goat Polyclonal Antibody
Type: Primary
Antigen: PSEN2 (presenilin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR