Anti-TUBA Mouse Monoclonal Antibody [clone: DM1A]

Supplier: Abcam
ABCAAB7291-100EA 508.36 CHF
ABCAAB7291-100
Anti-TUBA Mouse Monoclonal Antibody [clone: DM1A]
Antibodies
Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone: DM1A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR