Anti-Protein A Goat Polyclonal Antibody HRP (Horseradish Peroxidase)

ABCAAB7245-1EA 672 CHF
ABCAAB7245-1
Anti-Protein A Goat Polyclonal Antibody HRP (Horseradish Peroxidase)
Type: Primary
Antigen: Protein A
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR