Anti-Protein A Goat Polyclonal Antibody

ABCAAB7243-10EA 672 CHF
ABCAAB7243-10
Anti-Protein A Goat Polyclonal Antibody
Type: Primary
Antigen: Protein A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR