Anti-Streptavidin Rabbit Polyclonal Antibody HRP (Horseradish Peroxidase)

ABCAAB7239-1EA 672 CHF
ABCAAB7239-1
Anti-Streptavidin Rabbit Polyclonal Antibody HRP (Horseradish Peroxidase)
Type: Primary
Antigen: Streptavidin
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR