Anti-NFKBIA Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB7217-50EA 549.86 CHF
ABCAAB7217-50
Anti-NFKBIA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NFKBIA (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR