Anti-GLI2 Rabbit Polyclonal Antibody

ABCAAB7195-100EA 741 CHF
ABCAAB7195-100
Anti-GLI2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: GLI2 (GLI family zinc finger 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR