Anti-IgG Donkey Polyclonal Antibody

Supplier: Abcam
ABCAAB7120-500EA 207.49 CHF
ABCAAB7120-500
Anti-IgG Donkey Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Donkey
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR