Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Abcam
ABCAAB7098-500EA 274.93 CHF
ABCAAB7098-500
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR