Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Abcam
ABCAAB7050-1EA 248.99 CHF
ABCAAB7050-1
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR