Anti-EPHA1 Goat polyclonal antibody

ABCAAB7036-50EA 785 CHF
ABCAAB7036-50
Anti-EPHA1 Goat polyclonal antibody
Type: Primary
Antigen: EPHA1 (EPH receptor A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR