Anti-FCGR3A Mouse monoclonal antibody Biotin [clone: LNK16]

ABCAAB6998-100EA 824 CHF
ABCAAB6998-100
Anti-FCGR3A Mouse monoclonal antibody Biotin [clone: LNK16]
Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: monoclonal
Clone: LNK16
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR