Anti-IgG Goat Polyclonal Antibody (Cy2®)

Supplier: Abcam
ABCAAB6960-500EA 274.93 CHF
ABCAAB6960-500
Anti-IgG Goat Polyclonal Antibody (Cy2®)
Antibodies
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: Cy2®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR