Goat Anti- IgG Polyclonal Antibody (AP (Alkaline Phosphatase))

ABCAAB6935-1EA 311 CHF
ABCAAB6935-1
Goat Anti- IgG Polyclonal Antibody (AP (Alkaline Phosphatase))
Type: Secondary
Antigen: IgG (Bovine) (bovine IgG)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR