Anti-IgG Donkey Polyclonal Antibody (Texas Red®)

Supplier: Abcam
ABCAAB6898-1EA 248.99 CHF
ABCAAB6898-1
Anti-IgG Donkey Polyclonal Antibody (Texas Red®)
Antibodies
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Donkey
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR