Anti-IgG Goat Polyclonal Antibody (Texas Red®)

Lieferant: Abcam
ABCAAB6875-1EA 257.56 CHF
ABCAAB6875-1
Anti-IgG Goat Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR