Anti-IgG Donkey Polyclonal Antibody (Biotin)

Supplier: Abcam
ABCAAB6819-1EA 248.99 CHF
ABCAAB6819-1
Anti-IgG Donkey Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR