Anti-IgG Chicken Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abcam
ABCAAB6815-1EA 0 CHF
ABCAAB6815-1
Anti-IgG Chicken Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Chicken
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR