Anti-IgG Sheep Polyclonal Antibody (Texas Red®)

Lieferant: Abcam
ABCAAB6806-1EA 257.56 CHF
ABCAAB6806-1
Anti-IgG Sheep Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR