Anti-IgG Sheep Polyclonal Antibody (Texas Red®)

Supplier: Abcam
ABCAAB6793-1EA 248.99 CHF
ABCAAB6793-1
Anti-IgG Sheep Polyclonal Antibody (Texas Red®)
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR