Rabbit Anti- IgG Polyclonal Antibody (Texas Red®)

ABCAAB6781-1EA 328 CHF
ABCAAB6781-1
Rabbit Anti- IgG Polyclonal Antibody (Texas Red®)
Type: Secondary
Antigen: IgG (Golden Syrian Hamster) (golden syrian hamster IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR