Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)
Lieferant: Abcam
ABCAAB6751-1500EA 257.56 CHF
ABCAAB6751-1500
Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now